DL-Serin 99%

Lieferant: Thermo Scientific

Synonyme: (±)-2-Amino-3-hydroxypropionsäure, H-DL-Ser-OH, (±)-Serin

132650250 132651000 132655000
ACRO132650250EA 24.3 EUR
ACRO132650250 ACRO132651000 ACRO132655000
DL-Serin 99%
DL-Serin
Formel: C₃H₇NO₃
Molecular Weight: 105,09 g/mol
Siedepunkt: 395 °C (1013 hPa)
Schmelzpunkt: 240 °C (Zersetzung)
Dichte: 1,53 g/cm³
Lagertemperatur: Raumtemperatur
MDL: MFCD00064223
CAS-Nummer: 302-84-1
EINECS: 206-130-6
Merck Index: 13,08534

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Loss on drying ≤0.3 % (105°C, 3 h)
Sulfated ash ≤0.1 %
UV transmittance ≥95 % (c=10, 2 N HCl, 430 nm) 1 cm cell
Titration with HClO4 98.5 to 101.5 %
Chloride (Cl) ≤200 ppm
Sulfate (SO4) ≤500 ppm
Ammonium (NH4) ≤200 ppm
Iron (Fe) ≤10 ppm
Heavy metals (as Pb) ≤10 ppm
Other amino acids ≤0.5 % (TLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR