Cell culture media, McCoy's 5A
Lieferant: PAN BIOTECH
P04-05500 P04-05610
PANBP04-05500EA 16.2 EUR
PANBP04-05500 PANBP04-05610
Cell culture media, McCoy's 5A
Zellkulturmedien
McCoy's 5A medium is a complete medium with all amino acids and vitamins.
  • Suitable for growing primary cultures
This group contains marrow cells, gingival cells, adrenal cells, spleen cells, lung cells, rat embryos and other cell types.