Calciumacetat xHydrat, reinst

Lieferant: Bernd Kraft

15208.3600 15208.5600 15208.2600 15208.6600
KRAF15208.3600EA 57.5 EUR
KRAF15208.3600 KRAF15208.5600 KRAF15208.2600 KRAF15208.6600
Calciumacetat xHydrat, reinst
Calciumacetat xHydrat
Formel: (H₃CCOO)₂Ca·nH₂O
Dichte: 1,5 g/cm³ (20 °C)
MDL: MFCD00150019
CAS-Nummer: 114460-21-8
EINECS: 200-540-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR