Europiumtriacetat Hydrat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Europium(III)acetat Hydrat, Essigsäure Europium(III)salz Hydrat
M-Clarity™ Qualitätsstufe = MQ100
325627-25G
325627-5G
SIAL325627-25GEA
350
EUR
SIAL325627-25G
SIAL325627-5G
Europiumtriacetat Hydrat, Sigma-Aldrich®
Europiumtriacetat Hydrat
Formel:
C₆H₉EuO₆·H₂O Molecular Weight: 347,11 g/mol |
MDL:
MFCD00150115 CAS-Nummer: 62667-64-5 EINECS: 214-670-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...