D(-)-Lysin Monohydrochlorid ≥98%

Lieferant: Thermo Scientific

Synonyme: H-D-Lys-OH.HCl, (R)-(-)-2,6-Diaminohexansäure Hydrochlorid, (R)-(-)-2,6-Diaminocapronsäure Monohydrochlorid, (R)-(-)-Lysin Monohydrochlorid, (R)-(-)-Lysin Hydrochlorid, (R)-(-)-2,6-Diaminocapronsäure Hydrochlorid, (R)-(-)-2,6-Diaminohexansäure Monohydrochlorid, D(-)-Lysin Hydrochlorid

L07710.03 L07710.06
L07710.03EA 35.5 EUR
L07710.03 L07710.06
D(-)-Lysin Monohydrochlorid ≥98%
D(-)-Lysin Monohydrochlorid

Purity: 98% CAS No.: 7274-88-6 Molecular Formula: C6H14N2O2.HCl Formula Weight: 182.65 Melting Point: ca 260° dec. Beilstein Reference: 4356907

Formel: C₆H₁₅ClN₂O₂
Molecular Weight: 182,65 g/mol
Schmelzpunkt: ∼ 260 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00012920
CAS-Nummer: 7274-88-6
EINECS: 230-691-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR