D(-)-Lysin Monohydrochlorid
Lieferant: BACHEM BIOCHEMICA
Synonyme:
H-D-Lys-OH.HCl, (R)-(-)-2,6-Diaminohexansäure Hydrochlorid, (R)-(-)-2,6-Diaminocapronsäure Monohydrochlorid, (R)-(-)-Lysin Monohydrochlorid, (R)-(-)-Lysin Hydrochlorid, (R)-(-)-2,6-Diaminocapronsäure Hydrochlorid, (R)-(-)-2,6-Diaminohexansäure Monohydrochlorid, D(-)-Lysin Hydrochlorid
4017512.0005
4017512.0025
4017512.0100
BACE4017512.0005EA
0
EUR
BACE4017512.0005
BACE4017512.0025
BACE4017512.0100
D(-)-Lysin Monohydrochlorid
D(-)-Lysin Monohydrochlorid
Formel:
C₆H₁₅ClN₂O₂ Molecular Weight: 182,65 g/mol Schmelzpunkt: 261…262 °C |
MDL:
MFCD00012920 CAS-Nummer: 7274-88-6 EINECS: 230-691-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...