L(+)-Lysin Monohydrochlorid, Sigma-Aldrich®
Lieferant: Merck
Synonyme: (S)-(+)-Lysin Monohydrochlorid, H-Lys-OH.HCl, (S)-(+)-2,6-Diaminohexansäure Hydrochlorid, (S)-(+)-2,6-Diaminocapronsäure Hydrochlorid, L(+)-Lysin Hydrochlorid, (S)-(+)-2,6-Diaminohexansäure Monohydrochlorid, (S)-(+)-2,6-Diaminocapronsäure Monohydrochlorid, (S)-(+)-Lysin Hydrochlorid
M-Clarity™ Qualitätsstufe = MQ200
L5626-100G L5626-1KG L5626-500G L5626-5KG
SIALL5626-100GEA 30.5 EUR
SIALL5626-100G SIALL5626-1KG SIALL5626-500G SIALL5626-5KG
L(+)-Lysin Monohydrochlorid, Sigma-Aldrich®
L(+)-Lysin Monohydrochlorid
Formel: H₂N(CH₂)₄CH(NH₂)CO₂H·HCl
Molecular Weight: 182.65 g/mol
Schmelzpunkt: 263…264 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00064564
CAS-Nummer: 657-27-2
EINECS: 211-519-9