2',4',6'-Trihydroxyacetophenon 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Phloroacetophenone, THAP, 2-Acetylphloroglucinol
139790250
139790050
139791000
ACRO139790250EA
137
EUR
ACRO139790250
ACRO139790050
ACRO139791000
2',4',6'-Trihydroxyacetophenon 98%
2',4',6'-Trihydroxyacetophenon
Formel:
C₈H₈O₄ Molecular Weight: 168,15 g/mol Siedepunkt: 394 °C (1013 hPa) Schmelzpunkt: 219…222 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149091 CAS-Nummer: 480-66-0 EINECS: 207-556-5 |
Spezifikation Testergebnisse
Appearance | Yellow to orange to tan Powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Melting point | 217 to 222 °C |
Water | ≤10 % (K.F.) |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...