2',4',6'-Trihydroxyacetophenon, Hyrdrat ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Phloroacetophenone, THAP, 2-Acetylphloroglucinol

L04156.06 L04156.14
L04156.06EA 46.3 EUR
L04156.06 L04156.14
2',4',6'-Trihydroxyacetophenon, Hyrdrat ≥98%
2',4',6'-Trihydroxyacetophenon

Purity: 98+% CAS No.: 480-66-0 Molecular Formula: C8H8O4.xH2O Formula Weight: 168.15(anhy) Melting Point: 218-223° Beilstein Reference: 1911197

Formel: C₈H₈O₄
Molecular Weight: 168,15 g/mol
Siedepunkt: 394 °C (1013 hPa)
Schmelzpunkt: 218…223 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00149091
CAS-Nummer: 480-66-0
EINECS: 207-556-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR