(R)-(-)-3-Chlor-1,2-propandiol 97%, ee 98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
(R)-(-)-3-Chlor-1,2-dihydroxypropan
L18555.06
L18555.14
L18555.06EA
71.7
EUR
L18555.06
L18555.14
(R)-(-)-3-Chlor-1,2-propandiol 97%, ee 98%
(R)-(-)-3-Chlor-1,2-propandiol
Purity: 97%
CAS No.: 57090-45-6
Molecular Formula: C3H7ClO2
Formula Weight: 110.54
Flash Point: 137°(279°F)
Refractive Index: 1.4800
Boiling Point: 213°
Beilstein Reference: 2202928
Optical Rotation: -0.9°(neat)
UN No.: 2689
MDL No.: MFCD00135169
Formel:
ClCH₂CHOHCH₂OH Molecular Weight: 110.54 g/mol Siedepunkt: 213 °C Dichte: 1.322 Flammpunkt: 137 °C (279 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00135169 CAS-Nummer: 57090-45-6 EINECS: 435-560-9 UN: 2689 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...