Hexachloroplatinsäure Hydrat ≥99,9% (Metall-Basis)
Lieferant: Thermo Scientific
Gefahr
Synonyme: Hydrogenhexachloroplatinat(IV) Hydrat, Dihydrogenhexachloroplatinat(IV) Hydrat
11051.03 11051.06 11051.14 11051.22
11051.03EA 115 EUR
11051.03 11051.06 11051.14 11051.22
Hexachloroplatinsäure Hydrat ≥99,9% (Metall-Basis)
Hexachloroplatinsäure Hydrat
Purity: 99.9% CAS No.: 18497-13-7 Molecular Formula: H2PtCl6.6H2O Formula Weight: 517.91 (409.82anhy) Physical Form: Crystalline Density: 2.430 Appearance: Red-brown Odour: Odourless Melting Point: 60° dec. Solubility: Soluble in water, alcohol, acetone, ethyl acetate and ether Merck Reference: 14,7526 UN No.: 2507 MDL No.: MFCD00149910
Catalysis, electroplating, photography, platinum mirrors, printing for etching of zinc, producing fine Colour in glass and porcelain, indelible ink, microscopy
Formel: H₂Cl₆Pt·H₂O
Molecular Weight: 427,83 g/mol
Schmelzpunkt: 60 °C (Zers.)
Dichte: 2,430
Lagertemperatur: Raumtemperatur
MDL: MFCD00149909
CAS-Nummer: 26023-84-7
EINECS: 607-848-6
UN: 2507
ADR: 8,III
Merck Index: 14,7526