Hexachloroplatinsäure Hexahydrat (37,50% Pt), Pulver ACS

Lieferant: MP Biomedicals
Gefahr

Synonyme: Hydrogenhexachloroplatinat(IV) Hexahydrat, Dihydrogenhexachloroplatinat(IV) Hexahydrat

0215250901 0215250905
ICNA0215250901EA 205 EUR
ICNA0215250901 ICNA0215250905
Hexachloroplatinsäure Hexahydrat (37,50% Pt), Pulver ACS
Hexachloroplatinsäure Hexahydrat

Chloroplatinic acid is used in potassium determination, purification of platinum and in catalysis.


Alkali & other salts (as sulphates) ≤0,05%


Chloroplatinic acid is used in potassium determination, purification of platinum and in catalysis.


Soluble in alcohol; very soluble in water.

Formel: H₂PtCl₆·6H₂O
Molecular Weight: 517,9 g/mol
Schmelzpunkt: 60 °C (Lit.)
Dichte: 2,434 g/cm³ (20 °C)
MDL: MFCD00149910
CAS-Nummer: 18497-13-7
EINECS: 241-010-7
UN: 2507
ADR: 8,III
Merck Index: 13,07609

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR