D(-)-Serin
Lieferant: FLUOROCHEM
Synonyme:
H-D-Ser-OH, (R)-(-)-2-Amino-3-hydroxypropionsäure, (R)-(-)-Serin
011887-100G
011887-25G
FLUO011887-100GEA
92.1
EUR
FLUO011887-100G
FLUO011887-25G
D(-)-Serin
D(-)-Serin
Formel:
C₃H₇NO₃ Molecular Weight: 105,09 g/mol Schmelzpunkt: 235 °C |
MDL:
MFCD00004269 CAS-Nummer: 312-84-5 EINECS: 206-229-4 Merck Index: 12,08604 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...