Lithiumiodid Hydrat ≥99,995% (Metall-Basis), Puratronic®
Lieferant: Thermo Scientific
Achtung
Synonyme: Lithium iodide hydrate
10738.14 10738.06
10738.14EA 243 EUR
10738.14 10738.06
Lithiumiodid Hydrat ≥99,995% (Metall-Basis), Puratronic®
Lithiumiodid Hydrat
Lithium iodide hydrate is used as a reagent for ester cleavage and decarboxylation and C-C bond forming reactions. It is also used as source of nucleophilic iodide, as a mild lewis acid, epoxide opening and as an additive for organometallic-mediated transformations.
Purity: 99.995% CAS No.: 85017-80-7 Molecular Formula: LiI.xH2O Formula Weight: 133.84(anhy) Physical Form: Crystalline Aggregates Sensitivity: Light and Air Sensitive and Hygroscopic Merck Reference: 14,5535 MDL No.: MFCD00149769
Formel: ILi·xH₂O
Molecular Weight: 133,84 g/mol (anhy)
Schmelzpunkt: 73 °C (-H₂O)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149769
CAS-Nummer: 85017-80-7
EINECS: 233-822-5
Merck Index: 14,5535