Lithiumiodid Hydrat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Lithium iodide hydrate
M-Clarity™ Qualitätsstufe = MQ100
223816-100G
223816-500G
SIAL223816-100GEA
90.9
EUR
SIAL223816-100G
SIAL223816-500G
Lithiumiodid Hydrat, Sigma-Aldrich®
Lithiumiodid Hydrat
Formel:
ILi·xH₂O Molecular Weight: 133,84 g/mol (anhy) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149769 CAS-Nummer: 17023-25-5 EINECS: 233-822-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...