Lithiumiodid Hydrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Lithium iodide hydrate

62550-100G 62550-500G
SIAL62550-100GEA 98.6 EUR
SIAL62550-100G SIAL62550-500G
Lithiumiodid Hydrat, Sigma-Aldrich®
Lithiumiodid Hydrat
Formel: ILi·xH₂O
Molecular Weight: 133,84 g/mol (anhy)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149769
CAS-Nummer: 17023-25-5
EINECS: 233-822-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR