L-Homoserin ≥99%
Lieferant: Thermo Fisher Scientific
Synonyme:
(S)-(-)-Homoserin, H-Hser-OH, (S)-(-)-2-Amino-4-hydroxybuttersäure
L12427.03
L12427.06
L12427.MD
L12427.03EA
112
EUR
L12427.03
L12427.06
L12427.MD
L-Homoserin ≥99%
L-Homoserin
Purity: 99%
CAS No.: 672-15-1
Molecular Formula: C4H9NO3
Formula Weight: 119.12
Melting Point: ca 203° dec.
Beilstein Reference: 1721681
Merck Reference: 14,4739
Optical Rotation: -9° (c=2 in water)
MDL No.: MFCD00063090
Formel:
C₄H₉NO₃ Molecular Weight: 119.12 g/mol Schmelzpunkt: ∼ 203 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00063090 CAS-Nummer: 672-15-1 EINECS: 211-590-6 Merck Index: 13,04760 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...