tri-Kaliumorthophosphat Monohydrat ≥95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Kaliumphosphat tribasisch Monohydrat, Kaliumphosphat Monohydrat
41586.36
41586.A3
41586.36EA
117
EUR
41586.36
41586.A3
tri-Kaliumorthophosphat Monohydrat ≥95%
tri-Kaliumorthophosphat Monohydrat
Purity: 95%
CAS No.: 27176-10-9
Molecular Formula: K3PO4.H2O
Formula Weight: 230.28 (212.27anhy)
Physical Form: Granular
Density: 2.564
MDL No.: MFCD00150410
Formel:
K₃O₄P·1H₂O Molecular Weight: 230.28 g/mol Dichte: 2.564 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00150410 CAS-Nummer: 27176-10-9 EINECS: 231-907-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...