L(-)-Cystin, Millipore®
Lieferant: Merck
Synonyme:
(R)-(-)-Cystin, (2R,2'R)-(-)-3,3'-Dithiobis(2-aminopropansäure), (H-Cys-OH)2
MQ100
1.02837.1000
1.02837.9010
1.02837.1000EA
630
EUR
1.02837.1000
1.02837.9010
L(-)-Cystin, Millipore®
L(-)-Cystin
Formel:
HOOCCH(NH₂)CH₂SSCH₂CH(NH₂)COOH Molecular Weight: 240.3 g/mol Siedepunkt: 428 Schmelzpunkt: 261…262 °C Dichte: 1.677 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064228 CAS-Nummer: 56-89-3 EINECS: 200-296-3 Merck Index: 13,02811 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...