Cell culture media, Ham's F12
Lieferant: PAN BIOTECH
P04-14550 P04-14559 P04-14500
PANBP04-14550EA 15.3 EUR
PANBP04-14550 PANBP04-14559 PANBP04-14500
Cell culture media, Ham's F12
Zellkulturmedien
Ham's F10 is an alternative to Ham's F12 and it was used primarily to culture CHO-cells. Today, Ham's F10 can be used with or without FBS for many different cell cultures. It is used for example for primary cells of rat and chicken, but also for human diploid cells.
  • Sterile
  • Store from 2 to 8 °C