D(+)-Cystin ≥98%

Lieferant: Thermo Scientific

Synonyme: (S)-(+)-Cystin, (H-D-Cys-OH)2, (2S,2'S)-(+)-3,3'-Dithiobis(2-aminopropansäure)

L13772.03 L13772.06
L13772.03EA 76.7 EUR
L13772.03 L13772.06
D(+)-Cystin ≥98%
D(+)-Cystin

Purity: 98% CAS No.: 349-46-2 Molecular Formula: C6H12N2O4S2 Formula Weight: 240.30 Odour: Odourless Melting Point: ca 250° dec. Beilstein Reference: 1728093 Merck Reference: 14,2782 Optical Rotation: +219° (c=1 in 1N HCl) MDL No.: MFCD00002610

Formel: HOOCCH(NH₂)CH₂SSCH₂CH(NH₂)COOH
Molecular Weight: 240,3 g/mol
Schmelzpunkt: ∼ 250 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00002610
CAS-Nummer: 349-46-2
EINECS: 206-486-2
Merck Index: 14,2782

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR