D(-)-N-Methylglucamin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (-)-N-Methyl-D-glucamin, (-)-1-Methylamino-1-desoxy-D-glucitol, Meglumin, (-)-1-Desoxy-1-(methylamino)-D-glucitol

M-Clarity™ Qualitätsstufe = MQ200

M2004-100G M2004-1KG M2004-500G
SIALM2004-100GEA 45 EUR
SIALM2004-100G SIALM2004-1KG SIALM2004-500G
D(-)-N-Methylglucamin, Sigma-Aldrich®
D(-)-N-Methylglucamin
Formel: H₃CNHCH₂(CH(OH))₄CH₂OH
Molecular Weight: 195,22 g/mol
Schmelzpunkt: 128…131 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004707
CAS-Nummer: 6284-40-8
EINECS: 228-506-9
Merck Index: 12,06154

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR