Magnesiumthiosulfat Hexahydrat ≥99%
Lieferant: Thermo Fisher Scientific
Achtung
Synonyme: Magnesiumhyposulfit Hexahydrat
46114.22 46114.36
46114.22EA 29 EUR
46114.22 46114.36
Magnesiumthiosulfat Hexahydrat ≥99%
Magnesiumthiosulfat Hexahydrat
Purity: 99% CAS No.: 13446-30-5 Molecular Formula: MgO3S2.6H2O Formula Weight: 244.53 (136.44anhy) Physical Form: Crystalline Density: 1.818 Appearance: White Melting Point: 170° (-3H2O) Solubility: Soluble in water. Insoluble in alcohol Merck Reference: 14,5694 MDL No.: MFCD00169871
Formel: MgO₃S₂·6H₂O
Schmelzpunkt: 170 °C (-3H₂O)
Dichte: 1,818
Lagertemperatur: Raumtemperatur
MDL: MFCD00169871
CAS-Nummer: 13446-30-5
EINECS: 233-340-5
Merck Index: 14,5694