Indium(III)nitrat Hydrat ≥99,999% (Metall-Basis), Puratronic®
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Indium trinitrate hydrate
10708.06
10708.14
10708.22
10708.06EA
52.4
EUR
10708.06
10708.14
10708.22
Indium(III)nitrat Hydrat ≥99,999% (Metall-Basis), Puratronic®
Indium(III)nitrat Hydrat
Purity: 99.999%
CAS No.: 207398-97-8
Molecular Formula: In(NO3)3.xH2O
Formula Weight: 300.83(anhy)
Physical Form: Crystalline Aggregates
Sensitivity: Hygroscopic
Appearance: Off-white
Melting Point: 100° -H2O
UN No.: 1477
MDL No.: MFCD00149742
Formel:
InN₃O₉·H₂O Molecular Weight: 318,85 g/mol Schmelzpunkt: 100 °C (-H₂O) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149742 CAS-Nummer: 207398-97-8 EINECS: 237-393-5 UN: 1477 ADR: 5.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...