Indium(III)nitrat Hydrat ≥99,99% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Indium trinitrate hydrate
40185.PG
40185.PF
40185.PGEA
0
EUR
40185.PG
40185.PF
Indium(III)nitrat Hydrat ≥99,99% (Metall-Basis)
Indium(III)nitrat Hydrat
Purity: 99.99%
CAS No.: 207398-97-8
Molecular Formula: InN3O9.xH2O
Formula Weight: 300.83(anhy)
Physical Form: Lump/Powder
Sensitivity: Hygroscopic
Appearance: Off-white
Melting Point: 100° -H2O
UN No.: 1477
MDL No.: MFCD00149742
Formel:
InN₃O₉·H₂O Molecular Weight: 318.85 g/mol Schmelzpunkt: 100 °C (-H₂O) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149742 CAS-Nummer: 207398-97-8 EINECS: 237-393-5 UN: 1477 ADR: 5.1 III |