Indium(III)nitrat Hydrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Indium trinitrate hydrate

M-Clarity™ Qualitätsstufe = MQ100

326127-10G 326127-50G
SIAL326127-10GEA 152 EUR
SIAL326127-10G SIAL326127-50G
Indium(III)nitrat Hydrat, Sigma-Aldrich®
Indium(III)nitrat Hydrat
Formel: InN₃O₉·H₂O
Molecular Weight: 318.85 g/mol
MDL: MFCD00149742
CAS-Nummer: 207398-97-8
EINECS: 237-393-5
UN: 1477
ADR: 5.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR