Indium(III) sulphate hydrate ≥99,999% (Metall-Basis), Puratronic®
Lieferant: Thermo Scientific
Synonyme:
Diindium trisulphate hydrate
10711.09
10711.18
10711.09EA
106
EUR
10711.09
10711.18
Indium(III) sulphate hydrate ≥99,999% (Metall-Basis), Puratronic®
Indium(III) sulphate hydrate
Purity: 99.999%
CAS No.: 13464-82-9
Molecular Formula: In2(SO4)3.xH2O (x≈6)
Formula Weight: 517.83(anhy)
Physical Form: Lump/Powder
Melting Point: 250° dec.
MDL No.: MFCD00149747
Formel:
O₁₂In₂S₃·xH₂O Molecular Weight: 517,83 g/mol (anhy) Schmelzpunkt: 250 °C (Zers.) Dichte: 3,44 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
CAS-Nummer:
13464-82-9 EINECS: 236-689-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...