Tetraethylammoniumfluorid Hydrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Tetrylammoniumfluorid Hydrat

M-Clarity™ Qualitätsstufe = MQ100

235911-25G 235911-5G
SIAL235911-25GEA 123 EUR
SIAL235911-25G SIAL235911-5G
Tetraethylammoniumfluorid Hydrat, Sigma-Aldrich®
Tetraethylammoniumfluorid Hydrat
Formel: C₈H₂₀FN·xH₂O
Molecular Weight: 149,25 g/mol (anhy)
MDL: MFCD00149990
CAS-Nummer: 98330-04-2
EINECS: 211-559-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR