Europium(III)bromid Hydrat ≥99,99% (REO, Basis der Oxide der Metalle der seltenen Erden), REacton®

Lieferant: Thermo Scientific
Achtung

13213.06 13213.14
13213.06EA 144 EUR
13213.06 13213.14
Europium(III)bromid Hydrat ≥99,99% (REO, Basis der Oxide der Metalle der seltenen Erden), REacton®
Europium(III)bromid Hydrat

Purity: 99.99% Molecular Formula: EuBr3.xH2O Formula Weight: 391.69(anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Odour: Odourless MDL No.: MFCD00049921

Formel: Br₃Eu·H₂O
Molecular Weight: 409,69 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00049921
CAS-Nummer: 560069-78-5
EINECS: 237-349-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR