Europiumtrinitrat Hydrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Europium (III) nitrate hydrate

M-Clarity™ Qualitätsstufe = MQ100

254061-10G 254061-1G
SIAL254061-10GEA 612 EUR
SIAL254061-10G SIAL254061-1G
Europiumtrinitrat Hydrat, Sigma-Aldrich®
Europiumtrinitrat Hydrat
Formel: EuN₃O₉·xH₂O
Molecular Weight: 337,98 g/mol (anhy)
MDL: MFCD00149698
CAS-Nummer: 100587-95-9
EINECS: 233-380-3
UN: 1477
ADR: 5.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR