Ytterbiumtrinitrat Pentahydrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Ytterbium(III)nitrat Pentahydrat

M-Clarity™ Qualitätsstufe = MQ100

209147-10G 209147-50G
SIAL209147-10GEA 91.6 EUR
SIAL209147-10G SIAL209147-50G
Ytterbiumtrinitrat Pentahydrat, Sigma-Aldrich®
Ytterbiumtrinitrat Pentahydrat
Formel: N₃O₉Yb·5H₂O
Molecular Weight: 449,13 g/mol
MDL: MFCD00149883
CAS-Nummer: 35725-34-9
EINECS: 237-384-6
UN: 1477
ADR: 5.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR