Ytterbiumtrinitrat Pentahydrat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Ytterbium(III)nitrat Pentahydrat
M-Clarity™ Qualitätsstufe = MQ100
217220-25G
217220-5G
SIAL217220-25GEA
447
EUR
SIAL217220-25G
SIAL217220-5G
Ytterbiumtrinitrat Pentahydrat, Sigma-Aldrich®
Ytterbiumtrinitrat Pentahydrat
Formel:
N₃O₉Yb·5H₂O Molecular Weight: 449,13 g/mol |
MDL:
MFCD00149883 CAS-Nummer: 35725-34-9 EINECS: 237-384-6 UN: 1477 ADR: 5.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...