Yttrium(III)acetylacetonat Hydrat ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)
Lieferant: Thermo Scientific
Synonyme:
Tris(2,4-pentandionato-O,O')yttrium(III) Hydrat, Yttrium(III)-2,4-pentandionat Hydrat, Tri(2,4-pentandionato-O,O')yttrium(III) Hydrat, Yttrium(III)-tris(2,4-pentandionato-O,O') Hydrat
13154.14
13154.22
13154.14EA
198
EUR
13154.14
13154.22
Yttrium(III)acetylacetonat Hydrat ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)
Yttrium(III)acetylacetonat Hydrat
Purity: 99.9%
CAS No.: 207801-29-4
Molecular Formula: C15H21O6Y.xH2O
Formula Weight: 386.24(anhy)
Physical Form: Powder
Formel:
C₁₅H₂₁O₆Y·H₂O Molecular Weight: 404,25 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013509 CAS-Nummer: 207801-29-4 EINECS: 239-607-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...