L(+)-Lysin Monohydrat, Millipore®
Lieferant: Merck
Synonyme:
K Monohydrat, (S)-(+)-2,6-Diaminohexansäure Monohydrat, H-Lys-OH.H2O, (S)-(+)-Lysin Monohydrat, (S)-(+)-2,6-Diaminocapronsäure Monohydrat, Lys Monohydrat
M-Clarity™ Qualitätsstufe = MQ100
1.12233.0100
1.12233.1000
1.12233.0100EA
144
EUR
1.12233.0100
1.12233.1000
L(+)-Lysin Monohydrat, Millipore®
L(+)-Lysin Monohydrat
Formel:
H₂N(CH₂)₄CH(NH₂)CO₂H·H₂O Molecular Weight: 164.2 g/mol Schmelzpunkt: 225 Lagertemperatur: Kühlschrank |
MDL:
MFCD00151035 CAS-Nummer: 39665-12-8 EINECS: 200-294-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...