Methylenblau Hydrat, hochrein biologische Färbemittel
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3,7-Bis(dimethylamino)phenothiaziniumchlorid Hydrat, Methylthioniniumchlorid Hydrat, N,N,N′,N′-Tetramethylthioninchlorid Hydrat
Sensitive stain for RNA and ribonuclease.
Methylene blue is used as a dye in different staining procedures viz. Wright's stain and Jenner's stain. It also serves as an indicator and medicine. Further, it is used to examine RNA and DNA under the microscope or in a gel. It is widely used as a redox indicator in analytical chemistry. It acts as a photosensitizer utilized to generate singlet oxygen in order to make organic peroxides by a Diels-Alder reaction. It is also used in the sulfide analysis. It plays an important role in aquaculture to treat fungal infections.
Formel:
C₁₆H₁₈ClN₃S·xH₂O Schmelzpunkt: ∼ 180 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00150006 CAS-Nummer: 122965-43-9 EINECS: 200-515-2 Merck Index: 14,6060 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...