Methylenblau Hydrat, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 3,7-Bis(dimethylamino)phenothiaziniumchlorid Hydrat, Methylthioniniumchlorid Hydrat, N,N,N′,N′-Tetramethylthioninchlorid Hydrat

M-Clarity™ Qualitätsstufe = MQ200

66720-100G 66720-25G 66720-500G
SIAL66720-100GEA 136 EUR
SIAL66720-100G SIAL66720-25G SIAL66720-500G
Methylenblau Hydrat, Sigma-Aldrich®
Methylenblau Hydrat
Formel: C₁₆H₁₈ClN₃S·xH₂O
Molecular Weight: 319,86 g/mol (anhy)
MDL: MFCD00150006
CAS-Nummer: 122965-43-9
EINECS: 200-515-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR