2-Bromheptan, Sigma-Aldrich®

Lieferant: Merck
Gefahr


M-Clarity™ Qualitätsstufe = MQ100

292389-100G 292389-25G
SIAL292389-100GEA 269 EUR
SIAL292389-100G SIAL292389-25G
2-Bromheptan, Sigma-Aldrich®
2-Bromheptan
Formel: H₃CBrCH(CH₂)₄CH₃
Molecular Weight: 179,1 g/mol
CAS-Nummer: 1974-04-5
EINECS: 217-824-3
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR