Cell culture medium, William's E
Lieferant: PAN BIOTECH
P04-29050
P04-29050S1
P04-29050S4
PANBP04-29050EA
24.9
EUR
PANBP04-29050
PANBP04-29050S1
PANBP04-29050S4
Cell culture medium, William's E
Zellkulturmedien
This sterile medium is used for long-term cultivation of adult rat liver epithelial cells.
- Stored at 2 to 8 °C