Acetaldehyde ammonia trimer ≥95,0% (durch titrimetrische Analyse)
Lieferant: TCI EUROPE
|
Achtung
|
TCIAA0201-500GEA
315
EUR
TCIAA0201-500G
TCIAA0201-25G
Acetaldehyde ammonia trimer ≥95,0% (durch titrimetrische Analyse)
Acetaldehyde ammonia trimer
Formel:
C₆H₂₁N₃O₃ Molecular Weight: 183,25 g/mol |
MDL:
MFCD00149559 CAS-Nummer: 58052-80-5 EINECS: 200-868-2 UN: 1841 ADR: 9,III |
Spezifikation Testergebnisse
Appearance | White to Orange to Green powder to crystal |
Purity (Neutralization titration) | min. 95.0 % |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...