2,3-Butandionmonoxim ≥99%

Lieferant: Thermo Fisher Scientific

Synonyme: Diacetylmonoxim

A14339.0E A14339.22 A14339.36
A14339.0EEA 1023 EUR
A14339.0E A14339.22 A14339.36
2,3-Butandionmonoxim ≥99%
2,3-Butandionmonoxim

Purity: 99% CAS No.: 57-71-6 Molecular Formula: C4H7NO2 Formula Weight: 101.11 Storage Temperature: 2°C to 8°C Melting Point: 74-77° Boiling Point: 185-186° Beilstein Reference: 605582 MDL No.: MFCD00002116

Formel: C₄H₇NO₂
Molecular Weight: 101.11 g/mol
Siedepunkt: 185…186 °C
Schmelzpunkt: 74…77 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00002116
CAS-Nummer: 57-71-6
EINECS: 200-348-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR