Tributyrin (Glycerintributyrat) ≥98%
Lieferant: Thermo Fisher Scientific
Synonyme:
Glycerintributyrat
A11830.30
A11830.18
A11830.0B
A11830.30EA
67.1
EUR
A11830.30
A11830.18
A11830.0B
Tributyrin (Glycerintributyrat) ≥98%
Tributyrin (Glycerintributyrat)
Purity: 98%
CAS No.: 60-01-5
Molecular Formula: C15H26O6
Formula Weight: 302.37
Flash Point: 174°(345°F)
Refractive Index: 1.4358
Melting Point: -75°
Boiling Point: 307-308°
Beilstein Reference: 1714746
Merck Reference: 14,9620
MDL No.: MFCD00009392
Formel:
C₁₅H₂₆O₆ Siedepunkt: 307…308 °C Schmelzpunkt: –75 °C Dichte: 1,032 Flammpunkt: 174 °C (345 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009392 CAS-Nummer: 60-01-5 EINECS: 200-451-5 Merck Index: 14,9620 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...