Octylbutyrat ≥97%
Lieferant: Thermo Fisher Scientific
Synonyme:
Butanoic acid octyl ester, Butyric acid octyl ester
B24978.18
B24978.30
B24978.18EA
27
EUR
B24978.18
B24978.30
Octylbutyrat ≥97%
Octylbutyrat
Purity: 97%
CAS No.: 110-39-4
Molecular Formula: CH3CH2CH2CO2(CH2)7CH3
Formula Weight: 172.27
MDL No.: MFCD00048939
Formel:
C₁₂H₂₄O₂ Molecular Weight: 200.32 g/mol Siedepunkt: 224 Schmelzpunkt: –56 °C Dichte: 0.87 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00048939 CAS-Nummer: 110-39-4 EINECS: 203-762-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...