Ethyldecanoat ≥99%
Lieferant: Thermo Fisher Scientific
Synonyme:
Ethylcaprat, Decansäureethylester, Caprinsäureethylester
A10657.0E
A10657.22
A10657.36
A10657.0EEA
287
EUR
A10657.0E
A10657.22
A10657.36
Ethyldecanoat ≥99%
Ethyldecanoat
Purity: 99%
CAS No.: 110-38-3
Molecular Formula: CH3(CH2)8CO2CH2CH3
Formula Weight: 200.32
Flash Point: 102°(216°F)
Refractive Index: 1.4250
Melting Point: -20°
Boiling Point: 250-252°
Beilstein Reference: 1762128
Merck Reference: 14,3776
MDL No.: MFCD00009581
Formel:
C₁₂H₂₄O₂ Molecular Weight: 200.32 g/mol Siedepunkt: 250…252 °C Schmelzpunkt: –20 °C Dichte: 0.862 Flammpunkt: 102 °C (216 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009581 CAS-Nummer: 110-38-3 EINECS: 203-761-9 Merck Index: 13,03811 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...