Ethylönantat ≥98%
Lieferant: Thermo Scientific
Achtung
|
Synonyme:
Ethylenantat, Heptansäureethylester, Önanthsäureethylester, Ethylheptanoat
A14729.22
A14729.36
A14729.22EA
27.9
EUR
A14729.22
A14729.36
Ethylönantat ≥98%
Ethylönantat
Purity: 98+%
CAS No.: 106-30-9
Molecular Formula: CH3(CH2)5CO2CH2CH3
Formula Weight: 158.24
Flash Point: 66°(151°F)
Refractive Index: 1.412
Melting Point: -66°
Boiling Point: 188-189°
Beilstein Reference: 1752311
Merck Reference: 14,3835
MDL No.: MFCD00009538
Formel:
C₉H₁₈O₂ Molecular Weight: 158,24 g/mol Siedepunkt: 188…189 °C Schmelzpunkt: –66 °C Dichte: 0,868 Flammpunkt: 66 °C (151 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009538 CAS-Nummer: 106-30-9 EINECS: 203-382-9 Merck Index: 14,3835 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...