Lithiumacetat Dihydrat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Acetic acid lithium salt
62393-100G-F
62393-500G-F
SIAL62393-100G-FEA
119
EUR
SIAL62393-100G-F
SIAL62393-500G-F
Lithiumacetat Dihydrat, Sigma-Aldrich®
Lithiumacetat Dihydrat
Formel:
H₃CCOOLi·2H₂O Molecular Weight: 102,02 g/mol Schmelzpunkt: 291 °C Dichte: 1,26 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00066949 CAS-Nummer: 6108-17-4 EINECS: 208-914-3 Merck Index: 13,05543 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...