7-Aminoönanthsäure

Lieferant: BACHEM BIOCHEMICA

Synonyme: ω-Aminoheptansäure, 7-Aminoheptansäure, ω-Aminoönanthsäure

4008344.0005 4008344.0001
BACE4008344.0005EA 0 EUR
BACE4008344.0005 BACE4008344.0001
7-Aminoönanthsäure
7-Aminoönanthsäure
Formel: C₇H₁₅NO₂
Molecular Weight: 145,2 g/mol
MDL: MFCD00008242
CAS-Nummer: 929-17-9
EINECS: 213-197-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR