tert-Butylcarbamat 98%
Lieferant: Thermo Scientific
Synonyme:
Carbamidsäure-tert-butylester
173450010
173450050
173450250
ACRO173450010EA
20.9
EUR
ACRO173450010
ACRO173450050
ACRO173450250
tert-Butylcarbamat 98%
tert-Butylcarbamat
Formel:
C₅H₁₁NO₂ Molecular Weight: 117,15 g/mol Schmelzpunkt: 105…109 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007962 CAS-Nummer: 4248-19-5 EINECS: 224-209-3 |
Spezifikation Testergebnisse
Appearance | White to light yellow Powder or crystals or needles |
Infrared spectrum | Conforms |
Melting point | 106 to 110 °C |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...