Silberdiethyldithiocarbamat ACS
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
N,N-Diethyldithiocarbamidsäure Silbersalz, (Diethyldithiocarbamato-S,S')silber, Diethyldithiocarbamidsäure Silbersalz, DETC
419330100
419330250
419331000
ACRO419330100EA
101
EUR
ACRO419330100
ACRO419330250
ACRO419331000
Silberdiethyldithiocarbamat ACS
Silberdiethyldithiocarbamat
Formel:
(C₂H₅)₂NCSSAg Molecular Weight: 256,14 g/mol Schmelzpunkt: 172…175 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00004929 CAS-Nummer: 1470-61-7 EINECS: 216-003-7 ADR: 9,III |
Spezifikation Testergebnisse
Appearance | Green to khaki Powder |
Infrared spectrum | Conforms |
Solubility | (in Pyridine) Passes test |
Suitability | (absorbance < 0.1) Passes test (for As determination) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...