Natriumtrifluoracetat
Lieferant: FLUOROCHEM
Synonyme:
Trifluoressigsäure Natriumsalz
003595-250G
003595-50G
003595-5000G
FLUO003595-250GEA
227
EUR
FLUO003595-250G
FLUO003595-50G
FLUO003595-5000G
Natriumtrifluoracetat
Natriumtrifluoracetat
Formel:
C₂F₃NaO₂ Molecular Weight: 136.01 g/mol Schmelzpunkt: 207…209 °C |
MDL:
MFCD00013217 CAS-Nummer: 2923-18-4 EINECS: 220-879-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...