2-[Bis(2-hydroxyethyl)amino]ethansulfonsäure, Millipore®

Lieferant: Merck Millipore Calbiochem

Synonyme: BES, N,N-Bis-(2-hydroxyethyl)-2-aminoethansulfonsäure

391334-100 391334-25
391334-100EA 290 EUR
391334-100 391334-25
2-[Bis(2-hydroxyethyl)amino]ethansulfonsäure, Millipore®
2-[Bis(2-hydroxyethyl)amino]ethansulfonsäure
Formel: C₆H₁₅NO₅S
Molecular Weight: 213.25 g/mol
Siedepunkt: 405
Schmelzpunkt: 152…156 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007533
CAS-Nummer: 10191-18-1
EINECS: 233-465-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR